missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-prolinal, 97%

Product Code. 11346799
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11346799 1 g 1g
11451140 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11346799

Brand: Thermo Scientific Alfa Aesar H26294.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 73365-02-3 | C10H17NO3 | 199.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73365-02-3
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD01321389
InChI Key YDBPZCVWPFMBDH-MRVPVSSYSA-N
Synonym n-boc-d-prolinal, n-tert-butoxycarbonyl-d-prolinal, r-tert-butyl 2-formylpyrrolidine-1-carboxylate, boc-d-prolinal, tert-butyl 2r-2-formylpyrrolidine-1-carboxylate, 1-pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, 2r, tert-butyl r-2-formylpyrrolidine-1-carboxylate, r-n-boc-prolinal, pubchem15956
PubChem CID 7009153
IUPAC Name tert-butyl (2R)-2-formylpyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1C=O

Specifications

Density 1.06
Boiling Point 228°C
Flash Point >110°C (230°F)
Refractive Index 1.4618
Quantity 1 g
Sensitivity Air sensitive
Optical Rotation +88° (c=1 in Chloroform)
Formula Weight 199.25
Percent Purity 97%
Chemical Name or Material N-Boc-D-prolinal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.