missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-prolinal, 97%

Product Code. 11451140
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11451140 5 g 5g
11346799 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451140 Supplier Thermo Scientific Alfa Aesar Supplier No. H26294.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 73365-02-3 | C10H17NO3 | 199.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73365-02-3
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD01321389
InChI Key YDBPZCVWPFMBDH-MRVPVSSYSA-N
Synonym n-boc-d-prolinal, n-tert-butoxycarbonyl-d-prolinal, r-tert-butyl 2-formylpyrrolidine-1-carboxylate, boc-d-prolinal, tert-butyl 2r-2-formylpyrrolidine-1-carboxylate, 1-pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, 2r, tert-butyl r-2-formylpyrrolidine-1-carboxylate, r-n-boc-prolinal, pubchem15956
PubChem CID 7009153
IUPAC Name tert-butyl (2R)-2-formylpyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1C=O

Specifications

Density 1.06
Boiling Point 228°C
Flash Point >110°C (230°F)
Refractive Index 1.4618
Quantity 5 g
Sensitivity Air sensitive
Optical Rotation +88° (c=1 in Chloroform)
Formula Weight 199.25
Percent Purity 97%
Chemical Name or Material N-Boc-D-prolinal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.