missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-proline methyl ester, 95%, Thermo Scientific™

Product Code. p-7041579
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15439168 250 mg 250mg
15419168 1 g 1g
15429168 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15439168 Supplier Thermo Scientific Alfa Aesar Supplier No. H62672.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Boc-D-proline methyl ester is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in DMSO.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73323-65-6
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD00190828
InChI Key WVDGSSCWFMSRHN-MRVPVSSYSA-N
Synonym boc-d-pro-ome, boc-d-proline methyl ester, r-1-tert-butyl 2-methyl pyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2r-pyrrolidine-1,2-dicarboxylate, n-boc-d-proline methyl ester, r-1-boc-pyrrolidine-2-carboxylic acid methyl ester, n-tert-butoxycarbonyl-d-proline methyl ester, o1-tert-butyl o2-methyl 2r-pyrrolidine-1,2-dicarboxylate, boc-d-proline-methyl ester, pubchem16716
PubChem CID 1277667
IUPAC Name 1-O-tert-butyl 2-O-methyl (2R)-pyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1C(=O)OC

Specifications

Quantity 250 mg
Solubility Information Soluble in DMSO.
Formula Weight 229.27
Percent Purity 95%
Chemical Name or Material N-Boc-D-proline methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.