missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-tert-leucine, 95%, Thermo Scientific Chemicals

Product Code. 15448988
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15448988

Brand: Thermo Scientific Alfa Aesar H62473.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124655-17-0
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.29
MDL Number MFCD00065575
InChI Key LRFZIPCTFBPFLX-JLDDOWRYNA-N
Synonym boc-d-tert-leucine, n-boc-d-tert-leucine, boc-d-tle-oh, r-n-tert-butoxycarbonyl-tert-leucine, r-2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-d-alpha-t-butylglycine, boc-tbu-d-gly-oh, boc-d-t-leu, d-valine, n-1,1-dimethylethoxy carbonyl-3-methyl, n-tert-butoxycarbonyl-d-tert-leucine
PubChem CID 7005057
SMILES CC(C)(C)OC(=O)N[C@@H](C(O)=O)C(C)(C)C

Specifications

IUPAC Name (2R)-3,3-dimethyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
Quantity 1 g
Formula Weight 231.29
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-D-tert-leucine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.