missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-DL-nipecotic acid, 97%, Thermo Scientific™

Product Code. 11378446
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378446 1 g 1g
11388446 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378446 Supplier Thermo Scientific Alfa Aesar Supplier No. L17478.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84358-12-3
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
MDL Number MFCD00673775,MFCD02179173
InChI Key NXILIHONWRXHFA-UHFFFAOYNA-N
Synonym 1-boc-piperidine-3-carboxylic acid, 1-tert-butoxycarbonyl piperidine-3-carboxylic acid, 1-boc-3-piperidinecarboxylic acid, n-boc-nipecotic acid, 1-tert-butoxycarbonyl-3-piperidinecarboxylic acid, boc-dl-nip-oh, n-boc-3-piperidinecarboxylic acid, n-boc-piperidine-3-carboxylic acid, 1-n-boc-piperidine-3-carboxylic acid, n-boc-dl-nipecotic acid
PubChem CID 2735647
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCC(C1)C(O)=O

Specifications

Melting Point 160°C to 162°C
Quantity 1 g
Beilstein 5539297
Formula Weight 229.28
Percent Purity 97%
Chemical Name or Material N-Boc-DL-nipecotic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.