missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-ethanolamine, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | N-Boc-ethanolamine |
| CAS | 26690-80-2 |
| Molecular Formula | C7 H15 N O3 |
| Quantity | 1 g |
| Synonym | Ethanolamine Impurity 1 |
| InChI Formula | InChI=1S/C7H15NO3/c1-7(2,3)11-6(10)8-4-5-9/h9H,4-5H2,1-3H3,(H,8,10) |
| SMILES | CC(C)(C)OC(=O)NCCO |
| IUPAC Name | tert-butyl N-(2-hydroxyethyl)carbamate |
| Molecular Weight (g/mol) | 161.2 |
| Formula Weight | 161.1052 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction