missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-glycine methyl ester, 97%

Product Code. 15475478
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
Product Code. Quantity unitSize
15475478 5 mL 5mL
15485478 25 mL 25mL
2 options
This item is not returnable. View return policy

Product Code. 15475478

Brand: Thermo Scientific Alfa Aesar H59234.AA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 31954-27-5 | C8H15NO4 | 189.211 g/mol

It is used as a pharmaceutical, chemical, paint and dye intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical, chemical, paint and dye intermediate.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31954-27-5
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.211
MDL Number MFCD00038267
InChI Key PHUZOEOLWIHIKH-UHFFFAOYSA-N
Synonym boc-glycine methyl ester, boc-gly-ome, n-tert-butoxycarbonyl glycine methyl ester, n-boc-glycine methyl ester, boc-glycinemethylester, methyl 2-tert-butoxycarbonyl amino acetate, methyl n-tert-butoxycarbonyl glycinate, glycine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl tert-butoxycarbonyl amino acetate
PubChem CID 637609
IUPAC Name methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
SMILES CC(C)(C)OC(=O)NCC(=O)OC

Specifications

Density 1.079
Boiling Point 190°C
Flash Point 109°C (228°F)
Linear Formula (CH3)3COCONHCH2CO2CH3
Refractive Index 1.437
Quantity 5 mL
Formula Weight 189.21
Percent Purity 97%
Chemical Name or Material N-Boc-glycine methyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.