missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-glycine N-succinimidyl ester, 98%, Thermo Scientific Chemicals

Product Code. 15469048
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15469048 5 g 5g
15479048 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469048 Supplier Thermo Scientific Alfa Aesar Supplier No. H62528.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3392-07-2

Specifications

Melting Point 165°C to 167°C
Molecular Formula C15H24N2O6
MDL Number MFCD00022585 MFCD00022585
Quantity 5 g
Beilstein 1549242
Sensitivity Hygroscopic
Synonym boc-ile-osu, boc-l-isoleucine hydroxysuccinimide ester, boc-l-isoleucine n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 2s,3s-2-tert-butoxycarbonyl amino-3-methylpentanoate, boc-lle-osu, boc-ile-osu hplc, n-alpha-t-butyloxycarbonyl-l-isoleucine succinimidyl ester, 1-n-1,1-dimethylethoxy carbonyl-l-isoleucyl oxy-2,5-pyrrolidinedione, 2,5-dioxopyrrolidin-1-yl 2s,3s-3-methyl-2-2-methylpropan-2-yl oxycarbonylamino pentanoate
InChI Key FATJLEZSGFVHQA-CABZTGNLSA-N
SMILES CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) (2R)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoate
Molecular Weight (g/mol) 328.37
PubChem CID 131632401
Formula Weight 272.25
Percent Purity 98%
Chemical Name or Material N-Boc-glycine N-succinimidyl ester
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.