missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-glycine N-succinimidyl ester, 98%, Thermo Scientific Chemicals

Product Code. 15479048
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15479048 25 g 25g
15469048 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15479048 Supplier Thermo Scientific Alfa Aesar Supplier No. H62528.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3392-07-2 | Molecular Formula: C11H16N2O6 | Molecular Weight: 272.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3392-07-2

Specifications

Melting Point 165°C to 167°C
Molecular Formula C11H16N2O6
MDL Number MFCD00042761
Quantity 25 g
Beilstein 1549242
Sensitivity Hygroscopic
Synonym boc-gly-osu, boc-glycine n-hydroxysuccinimide ester, boc-glycine-n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino acetate, tert-butyl 2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl carbamate, bocglyosu, 2,5-dioxopyrrolidin-1-yl n-tert-butoxycarbonyl glycinate
InChI Key LJCWRJYVPJJTMB-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NCC(=O)ON1C(=O)CCC1=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
Molecular Weight (g/mol) 272.26
PubChem CID 76926
Formula Weight 272.25
Percent Purity 98%
Chemical Name or Material N-Boc-glycine N-succinimidyl ester
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.