missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-guanidine, 95%

Product Code. 15459778
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459778 25 g 25g
15439778 1 g 1g
15449778 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15459778 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur H63206.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 219511-71-4 | C6H13N3O2 | 159.189 g/mol

N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in most alcohols, esters (e.g., ethyl acetate), ketones (e.g., acetone), acetonitrile, and dichloromethane. Not soluble in diethyl ether and hexane.

Notes
Moisture, heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 219511-71-4
Molecular Formula C6H13N3O2
Molecular Weight (g/mol) 159.189
MDL Number MFCD20547465
InChI Key UMOZLQVSOVNSCA-UHFFFAOYSA-N
Synonym boc-guanidine, n-boc-guanidine, 1-tert-butoxycarbonyl guanidine, tert-butyl n-carbamimidoylcarbamate, boc guanidine, 1-boc-guanidine, tert-butyl n-diaminomethylidene carbamate, tert-butoxycarbonylguanidine, acmc-209x9z
PubChem CID 11378649
IUPAC Name tert-butyl N-(diaminomethylidene)carbamate
SMILES CC(C)(C)OC(=O)N=C(N)N

Spécification

Melting Point 170°C to 175°C
Color White
Linear Formula (CH3)3OCONHC(=NH)NH2
Quantity 25 g
Sensitivity Moisture sensitive
Solubility Information Soluble in most alcohols,esters (e.g.,ethyl acetate),ketones (e.g.,acetone),acetonitrile,and dichloromethane. Not soluble in diethyl ether and hexane.
Formula Weight 159.19
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-guanidine

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.