missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-hydroxylamine, 98+%

Product Code. 11464143
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464143 1 g 1g
11474143 5 g 5g
11484143 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464143 Supplier Thermo Scientific Alfa Aesar Supplier No. A10349.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 36016-38-3 | C5H11NO3 | 133.15 g/mol

N-Boc-hydroxylamine is used in the preparation of azridines by cycloaddition of azides with nitroso Diels-Alder adducts. It acts as a reagent for the synthesis of hydroxylamine derivatives t-butyl-N-(acyloxy)carbamates and N,O-diacylated N-hydroxyarylsulfonamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-hydroxylamine is used in the preparation of azridines by cycloaddition of azides with nitroso Diels-Alder adducts. It acts as a reagent for the synthesis of hydroxylamine derivatives t-butyl-N-(acyloxy)carbamates and N,O-diacylated N-hydroxyarylsulfonamides.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong bases, reducing agents and alkali metals. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36016-38-3
Molecular Formula C5H11NO3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00002107
InChI Key DRDVJQOGFWAVLH-UHFFFAOYSA-N
Synonym n-boc-hydroxylamine, n-tert-butoxycarbonylhydroxylamine, tert-butyl hydroxycarbamate, boc-amino alcohol, carbamic acid, hydroxy-, 1,1-dimethylethyl ester, n-hydroxycarbamic acid tert-butyl ester, tert-butyl-n-hydroxycarbamate, n-tert-boc-hydroxylamine, t-butyl n-hydroxycarbamate, n-tert-butyloxycarbonylhydroxylamine
PubChem CID 97534
IUPAC Name tert-butyl N-hydroxycarbamate
SMILES CC(C)(C)OC(=O)NO

Specifications

Melting Point 55°C to 58°C
Quantity 1 g
Beilstein 1756546
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 133.15
Percent Purity ≥98%
Chemical Name or Material N-Boc-hydroxylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.