missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-hydroxylamine, TRC

Product Code. 30348500
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348500 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348500 Supplier Toronto Research Chemicals Supplier No. TRCB6564705G
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Boc-hydroxylamine
CAS 36016-38-3
Molecular Formula C5H11NO3
Quantity 5 g
Synonym N-Hydroxycarbamic Acid 1,1-Dimethylethyl Ester, Hydroxycarbamic Acid 1,1-Dimethylethyl Ester, Hydroxycarbamic Acid tert-Butyl Ester, 1,1-Dimethylethyl N-Hydroxycarbamate, N-(tert-Butoxycarbonyl)hydroxylamine, N-(tert-Butyloxycarbonyl)hydroxylamine, N-Boc-hydroxylamine, N-Hydroxycarbamic Acid tert-Butyl Ester, N-t-Butyloxycarbonyl Hydroxylamine, NSC 131086, NSC 144620, tert-Butoxycarbonylhydroxylamine, tert-Butoxyhydroxamic Acid, tert-Butyl N-Hydroxycarbamate, tert-Butyl Hydroxycarbamate
InChI Formula InChI=1S/C5H11NO3/c1-5(2,3)9-4(7)6-8/h8H,1-3H3,(H,6,7)
SMILES CC(C)(C)OC(=O)NO
IUPAC Name tert-butyl N-hydroxycarbamate
Molecular Weight (g/mol) 133.15
Formula Weight 133.0739
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.