missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-alanyl-L-alanine, TRC

Product Code. 30179391
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179391 250 mg 250mg
30179188 500 mg 500mg
30179121 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179391 Supplier Toronto Research Chemicals Supplier No. TRCB600460250MG
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27317-69-7
Synonym NSC 334362∣N-tert-ButoxycarbonylalanylalanineTert-Butoxycarbonyl-L-alanyl-L-alanine∣N-Boc-L-alanyl-L-alanine

Specifications

Format Neat
Molecular Weight (g/mol) 260.29
Quantity 250 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.