missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-glutamic acid 5-benzyl ester, 98%

Product Code. 15433438
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15433438

Brand: Thermo Scientific Alfa Aesar H56879.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13574-13-5 | C17H22NO6 | 336.37 g/mol

N-Boc-L-glutamic acid 5-benzyl ester is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-glutamic acid 5-benzyl ester is used as a pharmaceutical intermediate.

Solubility
Soluble in N,N- Dimethyl formamide.

Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13574-13-5
Molecular Formula C17H22NO6
Molecular Weight (g/mol) 336.37
MDL Number MFCD00065569
InChI Key AJDUMMXHVCMISJ-ZDUSSCGKSA-M
Synonym boc-glu obzl-oh, boc-l-glutamic acid 5-benzyl ester, boc-l-glutamic acid 5-benzylester, 5-benzyl n-boc-l-glutamate, boc-l-glu obzl-oh, n-alpha-tert-boc-l-glutamic-gamma-benzyl ester, 5-benzyl n-tert-butoxycarbonyl-l-glutamate, boc-l-glutamic acid-5-benzyl ester, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, 5-phenylmethyl ester, n-boc-l-glutamic acid 5-benzyl ester
PubChem CID 83589
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CCC(=O)OCC1=CC=CC=C1)C([O-])=O

Specifications

Melting Point 69°C to 71°C
Quantity 5 g
Beilstein 2226572
Solubility Information Soluble in N,N- Dimethyl formamide.
Optical Rotation −5.5° (c=1 in Acetic Acid)
Formula Weight 337.37
Percent Purity 98%
Chemical Name or Material N-Boc-L-glutamic acid 5-benzyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.