missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-glutamic acid 5-methyl ester, 95%

Product Code. 15449018
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449018 5 g 5g
15459018 250 mg 250mg
15439018 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15449018 Supplier Thermo Scientific Alfa Aesar Supplier No. H62496.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 45214-91-3 | C11H19NO6 | 261.274 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 45214-91-3
Molecular Formula C11H19NO6
Molecular Weight (g/mol) 261.274
MDL Number MFCD00190794
InChI Key OHYMUFVCRVPMEY-ZETCQYMHSA-N
Synonym boc-glu ome-oh, s-2-tert-butoxycarbonyl amino-5-methoxy-5-oxopentanoic acid, boc-l-glutamic acid gamma-methyl ester, s-2-tert-butoxycarbonylamino-5-methoxy-5-oxopentanoic acid, 2s-2-tert-butoxycarbonyl amino-5-methoxy-5-oxopentanoic acid, pubchem18618, boc-l-glutamic acid methyl ester, n-boc-l-glutamic acid 5-methyl ester, n-t-butoxycarbonyl-l-glutamic acid 5-methyl ester, n-tert-butoxycarbonylglutamic acid ?-methyl ester
PubChem CID 7018756
IUPAC Name (2S)-5-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC(=O)OC)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 261.27
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-L-glutamic acid 5-methyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.