missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-BOC-L-Histidine, 99+%

Product Code. 10607684 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10607684 25 g Glass Bottle 25g
10127803 1 g Glass Bottle 1g
10530024 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10607684 Supplier Thermo Scientific Acros Supplier No. 275590250
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 17791-52-5 | C11H17N3O4 | 255.274 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17791-52-5
Molecular Formula C11H17N3O4
Molecular Weight (g/mol) 255.274
InChI Key AYMLQYFMYHISQO-QMMMGPOBSA-N
Synonym boc-his-oh, n-boc-l-histidine, boc-l-histidine, tert-butyloxycarbonyl-l-histidine, nalpha-tert-butoxycarbonyl-l-histidine, nalpha-boc-l-histidine, boc-l-his-oh, n-tert-butoxycarbonyl-l-histidine, l-histidine, n-1,1-dimethylethoxy carbonyl
PubChem CID 87308
IUPAC Name (2S)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Specific Rotation +24.5° to +26.5° (20°C, 589nm) (c=1, CH3OH)
Specific Rotation Condition +25.5° (25°C c=1,methanol)
Quantity 25 g
Formula Weight 255.27
Percent Purity ≥99.0%
Physical Form Powder, Crystals and/or Chunks
Chemical Name or Material Nα-BOC-L-Histidine, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.