missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-phenylalanine, 99%

Product Code. 11498636
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498636 25 g 25g
11488636 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11498636 Supplier Thermo Scientific Alfa Aesar Supplier No. A16017.14
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13734-34-4 | C14H19NO4 | 265.31 g/mol

N-Boc-L-phenylalanine is a derivative of Phenylalanine used in enantioselective hydrolysis of amino acid esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-phenylalanine is a derivative of Phenylalanine used in enantioselective hydrolysis of amino acid esters.

Solubility
Soluble in methanol, dichloromethane, dimethylformamide and N-methyl-2-pyrrolidone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13734-34-4
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00002663
InChI Key ZYJPUMXJBDHSIF-NSHDSACASA-N
Synonym boc-phe-oh, boc-l-phenylalanine, boc-phenylalanine, n-tert-butoxycarbonyl-l-phenylalanine, n-boc-l-phenylalanine, n-t-boc-l-phenylalanine, n-alpha-t-boc-l-phenylalanine, n-t-butyloxycarbonyl-l-phenylalanine, l-phenylalanine, n-1,1-dimethylethoxy carbonyl, boc-phe
PubChem CID 77857
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 84°C to 88°C
Quantity 25 g
Beilstein 2219729
Solubility Information Soluble in methanol,dichloromethane,dimethylformamide and N-methyl-2-pyrrolidone.
Optical Rotation +25° (c=1 in Ethanol)
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
Formula Weight 265.31
Percent Purity 99%
Chemical Name or Material N-Boc-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.