missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-phenylglycine, 99%

Product Code. 11359395
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11359395 1 g 1g
11369395 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11359395

Brand: Thermo Scientific Alfa Aesar L18541.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2900-27-8 | C13H17NO4 | 251.282 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2900-27-8
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.282
MDL Number MFCD00065588
InChI Key HOBFSNNENNQQIU-JTQLQIEISA-N
Synonym boc-phg-oh, boc-l-phenylglycine, s-2-tert-butoxycarbonyl amino-2-phenylacetic acid, boc-l-alpha-phenylglycine, boc-l-phg-oh, boc-l-a-phenylglycine, n-boc-l-alpha-phenylglycine, n-boc-l-2-phenylglycine, s-boc-l-phenylglycine, n-boc-l-phenylglycine
PubChem CID 11010409
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-2-phenylacetic acid
SMILES CC(C)(C)OC(=O)NC(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 88°C to 90°C
Beilstein 3592362
Quantity 1 g
Formula Weight 251.28
Optical Rotation +141° (c=1 in Ethanol)
Percent Purity 99%
Chemical Name or Material N-Boc-L-phenylglycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.