missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-proline, 99%, Thermo Scientific Chemicals

Product Code. 11404784
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404784 5 g 5g
11414784 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404784 Supplier Thermo Scientific Alfa Aesar Supplier No. A13744.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-Boc-L-proline is used as an intermediate in organic synthesis. It is also used to prepare daclatasvir, which inhibits the hepatitis C virus (HCV) non-structural 5A (NS5A) protein. Further, it is used as a drug in the treatment of hepatitis C virus (HCV).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-proline is used as an intermediate in organic synthesis. It is also used to prepare daclatasvir, which inhibits the hepatitis C virus (HCV) non-structural 5A (NS5A) protein. Further, it is used as a drug in the treatment of hepatitis C virus (HCV).

Solubility
Soluble in acetic acid.

Notes
Moisture and light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15761-39-4
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD00037324
InChI Key ZQEBQGAAWMOMAI-ZETCQYMHSA-N
Synonym boc-l-proline, boc-pro-oh, n-boc-l-proline, n-tert-butoxycarbonyl-l-proline, boc-l-pro-oh, s-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butyloxycarbonyl-l-proline, tert-butoxycarbonyl-l-proline, 1-tert-butoxycarbonyl-l-proline, boc-l-pro
PubChem CID 85083
IUPAC Name (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCC1C(=O)O

Specifications

Melting Point 132°C to 136°C
Quantity 5 g
Beilstein 15828
Solubility Information Soluble in acetic acid.
Optical Rotation −61° (c=1 in Acetic Acid)
Formula Weight 215.25
Percent Purity 99%
Chemical Name or Material N-Boc-L-proline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.