missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-Threonine methyl ester, TRC

Product Code. 30335407
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335407 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335407 Supplier Toronto Research Chemicals Supplier No. TRCB61928510G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Boc-L-Threonine methyl ester
CAS 79479-07-5
Molecular Formula C10H19NO5
Quantity 10 g
Synonym N-(tert-Butoxycarbonyl)threonine methyl ester, N-[(1,1-dimethylethoxy)carbonyl]-L-Threonine methyl ester, Boc-Thr-OMe
InChI Formula InChI=1S/C10H19NO5/c1-6(12)7(8(13)15-5)11-9(14)16-10(2,3)4/h6-7,12H,1-5H3,(H,11,14)/t6-,7+/m1/s1
SMILES COC(=O)[C@@H](NC(=O)OC(C)(C)C)[C@@H](C)O
IUPAC Name methyl (2S,3R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoate
Molecular Weight (g/mol) 233.26
Formula Weight 233.1263
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.