missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-p-phenylenediamine, TRC

Product Code. 30340761
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30340761 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340761 Supplier Toronto Research Chemicals Supplier No. TRCB66172525G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Boc-p-phenylenediamine
CAS 71026-66-9
Molecular Formula C11 H16 N2 O2
Quantity 25 g
Synonym (4-Aminophenyl)carbamic Acid 1,1-Dimethylethyl Ester, (4-Aminophenyl)carbamic Acid tert-Butyl Ester, 1,1-Dimethylethyl (4-Aminophenyl)carbamate, 4-(tert-Butoxycarbonylamino)aniline, 4-Amino-N-tert-butoxycarbonylaniline, 4-t-Butoxycarbonylaminoaniline, 4-tert-Butoxycarbonylaminophenylamine, N-(4-Aminophenyl)carbamic Acid 1,1-dimethylethyl Ester, N-(t-Butoxycarbonyl)-4-aminoaniline, N-(tert-Butoxycarbonyl)-1,4-PhenylenediamineN-(tert-Butoxycarbonyl)-p-phenylenediamine, O-(1,1-Dimethylethyl) N-(4-Aminophenyl)carbamate, tert-Butyl (4-Aminophenyl)carbamate, tert-Butyl N-(4-Anilino)carbamate
InChI Formula InChI=1S/C11H16N2O2/c1-11(2,3)15-10(14)13-9-6-4-8(12)5-7-9/h4-7H,12H2,1-3H3,(H,13,14)
SMILES CC(C)(C)OC(=O)Nc1ccc(N)cc1
IUPAC Name tert-butyl N-(4-aminophenyl)carbamate
Molecular Weight (g/mol) 208.257
Formula Weight 208.121
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.