missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-4-Piperidinecarboxylic acid, 98%

Product Code. p-3586458 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10752285 1 g Glass bottle 1g
10529233 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10752285 Supplier Thermo Scientific Acros Supplier No. 384870010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 84358-13-4 | C11H19NO4 | 229.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84358-13-4
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
MDL Number MFCD00076999
InChI Key JWOHBPPVVDQMKB-UHFFFAOYSA-N
Synonym n-boc-isonipecotic acid, 1-tert-butoxycarbonyl piperidine-4-carboxylic acid, n-boc-piperidine-4-carboxylic acid, 1-boc-piperidine-4-carboxylic acid, boc-inp-oh, 1-boc-isonipecotic acid, 1-tert-butoxycarbonyl-4-piperidinecarboxylic acid, boc-isonipecotic acid, 1-boc-4-piperidinecarboxylic acid, 1-tert-butoxy carbonyl piperidine-4-carboxylic acid
PubChem CID 392871
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-4-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 149°C to 153°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 229.27
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-4-Piperidinecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-Piperidine-4-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.