missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-Pyrrolidine, 97%

Product Code. 10366254 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10366254 5 g Glass bottle 5g
10264864 1 g Glass bottle 1g
10613025 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10366254 Supplier Thermo Scientific Acros Supplier No. 425820050
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket
This item is not returnable. View return policy

CAS: 86953-79-9 | C9H17NO2 | 171.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86953-79-9
Molecular Formula C9H17NO2
Molecular Weight (g/mol) 171.24
MDL Number MFCD00216581
InChI Key LPQZERIRKRYGGM-UHFFFAOYSA-N
Synonym 1-boc-pyrrolidine, n-boc-pyrrolidine, pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 1-pyrrolidinecarboxylate, 1-tert-butoxycarbonyl pyrrolidine, 1-pyrrolidinecarboxylic acid tert-butyl ester, boc-pyrrolidine, acmc-20aivt, pubchem9422, 1-boc-3-hydropyrrolidine
PubChem CID 643455
IUPAC Name tert-butyl pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1

Specifications

Density 0.9770g/mL
Boiling Point 80°C (0.2 mmHg)
Flash Point 86°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 0.977
Formula Weight 171.24
Percent Purity 97%
Chemical Name or Material N-BOC-Pyrrolidine
compliance-icons
Product Identifier
  • N-BOC-Pyrrolidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.