missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-thiourea, 97%

Product Code. p-7055104
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420379 250 mg 250mg
15400379 1 g 1g
15410379 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15420379 Supplier Thermo Scientific Alfa Aesar Supplier No. H63599.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 268551-65-1 | C6H12N2O2S | 176.23 g/mol

N-Boc-thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 268551-65-1
Molecular Formula C6H12N2O2S
Molecular Weight (g/mol) 176.23
MDL Number MFCD04039987
InChI Key JKXQTODLIZBUDE-UHFFFAOYSA-N
Synonym n-boc-thiourea, n-tert-butoxycarbonylthiourea, n-boc thiourea, mono-boc-thiourea, acmc-20ak5u, n-tert-butoxycarbonyl-thiourea, n-tert.butoxycarbonyl thiourea, 1-tert-butoxycarbonyl thiourea, tert-butyl carbamothioylcarbamate
PubChem CID 4095660
IUPAC Name tert-butyl N-carbamothioylcarbamate
SMILES CC(C)(C)OC(=O)NC(N)=S

Specifications

Melting Point 142°C (decomposition)
Color White
Quantity 250 mg
Formula Weight 176
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-Boc-thiourea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.