missing translation for 'onlineSavingsMsg'
Learn More

N-Bromophthalimide, 98+%

Product Code. 11454683
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11454683

Brand: Thermo Scientific Alfa Aesar A11601.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2439-85-2 | C8H4BrNO2 | 226.029 g/mol

Brominating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2439-85-2
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.029
MDL Number MFCD00005888
InChI Key MARXMDRWROUXMD-UHFFFAOYSA-N
PubChem CID 75542
IUPAC Name 2-bromoisoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)Br

Specifications

Melting Point 204°C to 208°C
Quantity 5 g
Beilstein 131211
Sensitivity Light and moisture sensitive
Synonym n-bromophthalimide, 2-bromoisoindoline-1,3-dione, 2-bromo-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 2-bromo, n-bromophtalimide, n-bromo phthalimide, acmc-1cjlt, 2-bromobenzo c azoline-1,3-dione, 1h-isoindole-1,3 2h-dione,2-bromo
Formula Weight 226.03
Percent Purity ≥98%
Chemical Name or Material N-Bromophthalimide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.