missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ n-Butoxyacetic acid, 95%

Product Code. 11429320
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429320 5 g 5g
11419320 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429320 Supplier Alfa Aesar™ Supplier No. H50210.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2516-93-0
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00067044
InChI Key AJQOASGWDCBKCJ-UHFFFAOYSA-N
Synonym butoxyacetic acid, n-butoxyacetic acid, acetic acid, butoxy, ccris 7195, acetic acid, 2-butoxy, 2-n-butoxy acetic acid, butoxyaceticacid, 3-oxaheptanoic acid, acmc-1cb1h, ajqoasgwdcbkcj-uhfffaoysa
PubChem CID 41958
IUPAC Name 2-butoxyacetic acid
SMILES CCCCOCC(=O)O

Specifications

Boiling Point 234°C
Quantity 5 g
Formula Weight 132.16
Percent Purity 95%
Chemical Name or Material n-Butoxyacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.