missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butyl 4-Hydroxybenzoate-2,3,5,6-d4, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | n-Butyl 4-Hydroxybenzoate-2,3,5,6-d4 |
| CAS | 1219798-67-0 |
| Molecular Formula | C11 D4 H10 O3 |
| Quantity | 10 mg |
| Synonym | Benzoic-2,3,5,6-d4 acid, 4-hydroxy-, butyl ester (ACI), Butyl 4-hydroxybenzoate-2,3,5,6-d4 (ACI), 4-Hydroxybenzoic-2,3,5,6-d4 acid butyl ester, n-Butyl 4-Hydroxybenzoate-2,3,5,6-d4, Butyl Parahydroxybenzoate-d4, Butylparaben-d4, Butyl Parahydroxybenzoate-D4 (4-hydroxybenzoic-2,3,5,6-D4) |
| InChI Formula | InChI=1S/C11H14O3/c1-2-3-8-14-11(13)9-4-6-10(12)7-5-9/h4-7,12H,2-3,8H2,1H3/i4D,5D,6D,7D |
| SMILES | [2H]c1c([2H])c(C(=O)OCCCC)c([2H])c([2H])c1O |
| IUPAC Name | butyl 2,3,5,6-tetradeuterio-4-hydroxybenzoate |
| Molecular Weight (g/mol) | 198.252 |
| Formula Weight | 198.119 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction