missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl acetate, 99.5+%, for electronic use (MOS), residue free

Product Code. 10717582 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10717582 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10717582 Supplier Thermo Scientific Acros Supplier No. 327850010
  • ONLINE EXCLUSIVE PRICE
    64.20 €
    62.30 € / 1 L
    Save 1.90 €
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123-86-4 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For electronic use
TRUSTED_SUSTAINABILITY

Specifications

CAS 123-86-4
Melting Point -78.0°C
Density 0.8800g/mL
Boiling Point 127.0°C
Flash Point 27°C
Assay Percent Range 99.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C6H12O2
Linear Formula CH3CO2(CH2)3CH3
Refractive Index 1.3930 to 1.3950
MDL Number MFCD00009445
Quantity 1 L
Beilstein 02, 130
Residue after Evaporation 0.001% max.
Merck Index 15, 1537
Specific Gravity 0.88
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
Solubility Information Solubility in water: 7g/L (20°C)
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILES CCCCOC(C)=O
IUPAC Name butyl acetate
Molecular Weight (g/mol) 116.16
PubChem CID 31272
ChEBI CHEBI:31328
Viscosity 0.69 mPa.s (25°C)
Formula Weight 116.16
Percent Purity 99.5+%
Grade For electronic use (MOS)
Physical Form Liquid
Chemical Name or Material n-Butyl acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.