missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl acetate, 99+%, extra pure

Product Code. 10224660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
Unit Size:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10224660 1 L Glass Bottle 1 L
10294700 2.5 L Glass Bottle 2.5 L
10227023 25 L Metal drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10224660 Supplier Thermo Scientific Acros Supplier No. 107750010
  • ONLINE EXCLUSIVE PRICE
    58.45 €
    53.70 € / 1 L
    Save 4.75 €
    8% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123-86-4 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -78.0°C
Density 0.8800g/mL
Boiling Point 127.0°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass Bottle
Linear Formula CH3CO2(CH2)3CH3
Refractive Index 1.3930 to 1.3950
Quantity 1 L
Beilstein 02, 130
Residue after Evaporation 0.005% max.
Merck Index 15, 1537
Specific Gravity 0.88
Solubility Information Solubility in water: 7g/L (20°C)
Viscosity 0.69 mPa.s (25°C)
Formula Weight 116.16
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material n-Butyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Butyl acetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.