missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl acetate,Solstice

Product Code. 16625883
Change view
Click to view available options
Quantity:
175 kg
Unit Size:
175kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16625883 175 kg 175kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16625883 Supplier Fluka Supplier No. 65530175KG
  • ONLINE EXCLUSIVE PRICE
    3495.00 €
    2582.68 € / 175kg
    Save 912.32 €
    26% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 123-86-4
Melting Point -78°C
Color Colorless
Density 0.881 g/cm3 (20°C)
Boiling Point 126.2°C (1013 hPa)
Flash Point 27°C
Molecular Formula C6H12O2
Quantity 175 kg
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILES CCCCOC(C)=O
IUPAC Name butyl acetate
Molecular Weight (g/mol) 116.16
Grade ACS General Use Solvents
Chemical Name or Material n-Butyl acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.