missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl acetate, Semiconductor Grade, 99% min

Product Code. 11313567
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313567 4 x 1 L 4 L
11333567 1 L 1 L
11323567 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11313567 Supplier Thermo Scientific Alfa Aesar Supplier No. 019395.M1
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123-86-4 | C6H12O2 | 116.16 g/mol

n-Butyl acetate is widely used as a solvent for cellulose nitrate, resins, polymers, oils and fats. It is used in paints, coatings, printing inks, aerosol sprays and cosmetics. It is used as a synthetic fruit flavoring agent in foods like candy, ice cream, cheeses and baked food items. In the pharmaceutical industry, it is used for extraction and as processing solvent. It is an ingredient of cleaners, essences and fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Melting Point -78°C
Color Colorless
Density 0.881 g/mL
pH 5
Boiling Point 124°C to 126°C
Flash Point 25°C (77°F)
Odor Fruit-like
Linear Formula CH3CO2(CH2)3CH3
Refractive Index 1.3945
Quantity 4 x 1 L
UN Number UN1123
Beilstein 1741921
Merck Index 14,1535
Solubility Information Miscible with ethanol,ethyl ether,acetone,chloroform and most hydrocarbons. Slightly miscible with water.
Formula Weight 116.16
Percent Purity ≤99%
Grade Semiconductor
Physical Form Liquid
Chemical Name or Material n-Butyl acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.