missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl acrylate, 98+%, stab. with up to 50ppm 4-methoxyphenol, Thermo Scientific Chemicals

Product Code. 11392166
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392166 500 mL 500mL
11382166 100 mL 100mL
11302176 2500 mL 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11392166 Supplier Thermo Scientific Alfa Aesar Supplier No. L03476.AP
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 141-32-2 | C7H12O2 | 128.171 g/mol

n-Butyl acrylate is used in polymer manufacturing, textile, leather finishing, formulating paints and adhesive. It is also used in the production of homopolymers and co-polymers like acrylic acid and its salts, esters, amides, methacrylates, acrylonitrile, maleates, vinyl acetate, vinyl chloride, vinylidene chloride, styrene, butadiene and unsaturated polyesters. Further, it is also used in making vitamin B1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Butyl acrylate is used in polymer manufacturing, textile, leather finishing, formulating paints and adhesive. It is also used in the production of homopolymers and co-polymers like acrylic acid and its salts, esters, amides, methacrylates, acrylonitrile, maleates, vinyl acetate, vinyl chloride, vinylidene chloride, styrene, butadiene and unsaturated polyesters. Further, it is also used in making vitamin B1.

Solubility
Miscible with ethanol, ethyl ether and acetone. Slightly miscible with water and carbon tetrachloride.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, catalytic metals, organotin catalysts, amines, peroxides and halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-32-2
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00009446
InChI Key CQEYYJKEWSMYFG-UHFFFAOYSA-N
Synonym butyl acrylate, n-butyl acrylate, acrylic acid butyl ester, n-butyl propenoate, 2-propenoic acid, butyl ester, butyl 2-propenoate, butylacrylate, acrylic acid, butyl ester, acrylic acid n-butyl ester, butylester kyseliny akrylove
PubChem CID 8846
ChEBI CHEBI:3245
IUPAC Name butyl prop-2-enoate
SMILES CCCCOC(=O)C=C

Specifications

Melting Point -64°C
Density 0.895
Boiling Point 145°C to 146°C
Flash Point 39°C (102°F)
Odor Fruit-like
Linear Formula CH2=CHCO2CH2CH2CH2CH3
Refractive Index 1.418
Quantity 500 mL
UN Number UN2348
Beilstein 1749970
Sensitivity Light sensitive
Merck Index 14,1539
Solubility Information Miscible with ethanol,ethyl ether and acetone. Slightly miscible with water and carbon tetrachloride.
Formula Weight 128.17
Percent Purity ≥98%
Chemical Name or Material n-Butyl acrylate, stabilized with up to 50ppm 4-methoxyphenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Butyl acrylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: n-Butyl acrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.