missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl disulfide, 97%

Product Code. 11935721 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11935721 500 g Glass bottle 500g
10539820 25 g Glass bottle 25g
10498050 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11935721 Proveedor Thermo Scientific Acros N.º de proveedor 107925000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 629-45-8 | C8H18S2 | 178.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 629-45-8
Molecular Formula C8H18S2
Molecular Weight (g/mol) 178.35
MDL Number MFCD00009467
InChI Key CUDSBWGCGSUXDB-UHFFFAOYSA-N
Synonym dibutyl disulfide, butyl disulfide, disulfide, dibutyl, n-butyl disulfide, di-n-butyl disulfide, dibutyl disulphide, 5,6-dithiadecane, 1-butyldisulfanyl butane, unii-z77ef35c0q, 1-butyldisulfanyl-butane
PubChem CID 12386
IUPAC Name 1-(butyldisulfanyl)butane
SMILES CCCCSSCCCC

Especificaciones

Color Brown to Colorless
Density 0.9380g/mL
Boiling Point 229.0°C to 233.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula [CH3(CH2)3]2S2
Refractive Index 1.4915 to 1.4935
Quantity 500 g
Beilstein 01,II,4
Specific Gravity 0.938
Solubility Information Solubility in water: immiscible
Formula Weight 178.35
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material n-Butyl disulfide
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • n-Butyl disulfide
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.