missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl heptafluorobutyrate, 97%, Thermo Scientific™

Product Code. 11470517
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11470517

Brand: Thermo Scientific Alfa Aesar B22400.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1559-07-5
Molecular Formula C8H9F7O2
Molecular Weight (g/mol) 270.147
MDL Number MFCD00039240
InChI Key YDXXJZKOOOJVEE-UHFFFAOYSA-N
Synonym n-butyl heptafluorobutyrate, butyric acid, heptafluoro-, butyl ester, butyl heptafluorobutanoate, heptafluorobutyric acid, n-butyl ester, n-butylheptafluorobutyrate, butyl perfluorobutanoat, butyl perfluorobutyrate, butylheptafluorobutyrate, butyl heptafluorobutyrate, n-butyl perfluoro-n-butyrate
PubChem CID 137079
IUPAC Name butyl 2,2,3,3,4,4,4-heptafluorobutanoate
SMILES CCCCOC(=O)C(C(C(F)(F)F)(F)F)(F)F

Specifications

Density 1.296
Boiling Point 131°C to 133°C
Linear Formula CF3CF2CF2CO2(CH2)3CH3
Refractive Index 1.327
Quantity 50 g
Beilstein 1798327
Formula Weight 270.15
Percent Purity 97%
Chemical Name or Material n-Butyl heptafluorobutyrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.