missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl nitrite, 95%

Product Code. 15432988
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15432988 500 g 500g
15422988 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15432988 Supplier Thermo Scientific Alfa Aesar Supplier No. H56501.36
 Restricted Item
Estimated Shipment: 24-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 544-16-1 | C4H9NO2 | 103.121 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-16-1
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00002058
InChI Key JQJPBYFTQAANLE-UHFFFAOYSA-N
Synonym 1-butyl nitrite, nitrous acid, butyl ester, n-butyl nitrite, nitrous acid, n-butyl ester, nitrous acid butyl ester, n-butylnitrite, butylnitrite, unii-44p8qg0f3t, ccris 838, n-c4h9ono
PubChem CID 10996
IUPAC Name butyl nitrite
SMILES CCCCON=O

Specifications

Density 0.882
Boiling Point 78°C
Flash Point −13°C (9°F)
Linear Formula CH3(CH2)3ONO
Refractive Index 1.376
Quantity 500 g
UN Number UN2351
Beilstein 1701036
Solubility Information Slightly soluble in water.
Formula Weight 103.12
Percent Purity 95%
Chemical Name or Material n-Butyl nitrite
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.