missing translation for 'onlineSavingsMsg'
Learn More

N-Butyl Nor Scopolamine Hydrochloride, TRC

Product Code. 30135340
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135340 250 mg 250mg
30007462 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135340 Supplier Toronto Research Chemicals Supplier No. TRCB693580250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22235-98-9
Molecular Formula C20 H27 N O4 . Cl H
Molecular Weight (g/mol) 381.89
Synonym Benzeneacetic acid, α-(hydroxymethyl)-, 9-butyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester, hydrochloride, [7(S)-(1α,2β,4β,5α,7β)]- (9CI), 1αH,5αH-Nortropan-3α-ol, 8-butyl-6β,7β-epoxy-, (-)-tropate (ester), hydrochloride (8CI), 3-Oxa-9-azatricyclo[3.3.1.02,4]nonane, benzeneacetic acid deriv., Tropic acid, 8-butyl-6β,7β-epoxy-1αH,5αH-nortropan-3α-yl ester, hydrochloride, (-)- (8CI), N-Butylnorhyoscine Hydrochloride, N-Butylnorscopolamine Hydrochloride, (1R,2R,4S,5S,7s)-9-Butyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-7-yl (2S)-3-hydroxy-2-phenylpropanoate Hydrochloride, Hyoscine Butylbromide Imp. E (EP) as Hydrochloride
IUPAC Name (9-butyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-7-yl) (2S)-3-hydroxy-2-phenylpropanoate;hydrochloride
SMILES Cl.CCCCN1[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO)c4ccccc4

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C20H27NO4.ClH/c1-2-3-9-21-16-10-14(11-17(21)19-18(16)25-19)24-20(23)15(12-22)13-7-5-4-6-8-13;/h4-8,14-19,22H,2-3,9-12H2,1H3;1H/t14-,15-,16-,17+,18-,19+;/m1./s1
Formula Weight 381.1707
Percent Purity >95%
Chemical Name or Material N-butyl Norscopolamine hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.