missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl phosphate, mixture of mono-n-butyl and di-n-butyl

Product Code. 11378037
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378037 250 g 250g
11368037 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378037 Supplier Thermo Scientific Alfa Aesar Supplier No. 022404.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52933-01-4 | C4H11O4P | 154.102 g/mol

n-Butyl phosphate is a solvent and plasticizer for cellulose esters such as nitrocellulose and cellulose acetate. It forms stable hydrophobic complexes with some metals, as an extractant. It is also used in industry are as a component of aircraft hydraulic fluid and as a solvent for extraction and purification of rare earth metals from their ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52933-01-4
Molecular Formula C4H11O4P
Molecular Weight (g/mol) 154.102
MDL Number MFCD00013947
InChI Key BNMJSBUIDQYHIN-UHFFFAOYSA-N
Synonym n-butylphosphate, monobutyl phosphate, phosphoric acid, monobutyl ester, butyl diacid phosphate, n-butyl acid phosphate, butyl dibasic acid phosphate, unii-0i9w48e53l, butyl phosphate, buo ho 2po, mono-n-butylphosphoric acid, butoxyphosphonic acid
PubChem CID 15389
IUPAC Name butyl dihydrogen phosphate
SMILES CCCCOP(=O)(O)O

Specifications

Melting Point -60°C
Color Colorless
Density 1.123 g/mL
Boiling Point >124°C (decomposition)
Flash Point 110°C (230°F)
Refractive Index 1.429
Quantity 250 g
UN Number UN1718
Solubility Information Soluble in ethanol,diethyl ether. Insoluble in water.
Physical Form Liquid
Chemical Name or Material n-Butyl phosphate, Mixture of mono-n-butyl and di-n-butyl
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.