missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl sulfide, 99%, pure

Product Code. 10122230 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10122230 500 mL Glass bottle 500mL
10090710 5 mL Glass bottle 5mL
10694752 100 mL Glass bottle 100mL
11945721 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10122230 Supplier Thermo Scientific Acros Supplier No. 108015000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 544-40-1 | C8H18S | 146.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-40-1
Molecular Formula C8H18S
Molecular Weight (g/mol) 146.29
MDL Number MFCD00009468
InChI Key HTIRHQRTDBPHNZ-UHFFFAOYSA-N
Synonym dibutyl sulfide, butyl sulfide, n-butyl sulfide, butylthiobutane, di-n-butyl sulfide, butane, 1,1'-thiobis, butyl monosulfide, dibutyl thioether, thiononane-5, n-dibutyl sulfide
PubChem CID 11002
SMILES CCCCSCCCC

Specifications

Melting Point -80.0°C
Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 188.0°C to 189.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula [CH3(CH2)3]2S
Refractive Index 1.4520 to 1.4540
Quantity 500 mL
Beilstein 01,IV,1559
Merck Index 15,1593
Specific Gravity 0.84
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether,soluble in most organic solvents
IUPAC Name 1-(butylsulfanyl)butane
Formula Weight 146.29
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material n-Butyl sulfide
Show More Show Less
compliance-icons
Product Identifier
  • n-Butyl sulfide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.