missing translation for 'onlineSavingsMsg'
Learn More

n-Butylbenzene, 99+%

Product Code. 10530171 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10530171 50 mL Glass bottle 50mL
10756131 250 mL Glass bottle 250mL
10185303 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10530171 Supplier Thermo Scientific Acros Supplier No. 107850500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 104-51-8 | C10H14 | 134.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-51-8
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00009463
InChI Key OCKPCBLVNKHBMX-UHFFFAOYSA-N
Synonym n-butylbenzene, 1-phenylbutane, benzene, butyl, 1-butylbenzene, butyl-benzene, butane, phenyl, unii-s8xz2901rz, butylbenzene, analytical standard, n-butyl benzene, n4b
PubChem CID 7705
ChEBI CHEBI:44194
IUPAC Name butylbenzene
SMILES CCCCC1=CC=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -88°C
Color Colorless
Density 0.8600g/mL
Boiling Point 183°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C6H5(CH2)3CH3
Refractive Index 1.4880 to 1.49
Quantity 50 mL
Beilstein 05,413
Merck Index 15,1551
Specific Gravity 0.86
Solubility Information Solubility in water: insoluble in water. Other solubilities: miscible with ethanol,ether and benzene
Formula Weight 134.22
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material n-Butylbenzene, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • n-Butylbenzene
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.