missing translation for 'onlineSavingsMsg'
Learn More

n-Butyldimethylchlorosilane, 97+%, Thermo Scientific™

Product Code. 10666650 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10666650 10 mL Glass bottle 10mL
10715842 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10666650 Supplier Thermo Scientific Acros Supplier No. 432260100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1000-50-6
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.72
MDL Number MFCD00015727
InChI Key MXOSTENCGSDMRE-UHFFFAOYSA-N
Synonym butylchlorodimethylsilane, butyldimethylchlorosilane, n-butyldimethylchlorosilane, butyldimethylsilyl chloride, 1-butyldimethylchlorosilane, silane, butylchlorodimethyl, butyl chloro dimethylsilane, n-butyl-dimethylchlorosilane, butyl-chloro-dimethyl-silane, n-butyldimethylsilyl chloride
PubChem CID 66079
IUPAC Name butyl-chloro-dimethylsilane
SMILES CCCC[Si](C)(C)Cl

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless
Density 0.8750g/mL
Boiling Point 138.0°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH3)2Si(CH2)3CH3
Refractive Index 1.4190 to 1.4210 (20°C, 589nm)
Quantity 10 mL
Specific Gravity 0.875
Solubility Information Solubility in water: insoluble,reacts
Formula Weight 150.72
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material n-Butyldimethylchlorosilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Butyldimethylchlorosilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.