missing translation for 'onlineSavingsMsg'
Learn More

n-Butylmagnesium chloride, 20 wt% solution in THF/Toluene, AcroSeal™

Product Code. 11915851 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11915851

Brand: Thermo Scientific Acros 331688000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 693-04-9 | C4H9ClMg | 116.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-04-9, 109-99-9, 108-88-3
Molecular Formula C4H9ClMg
Molecular Weight (g/mol) 116.87
MDL Number MFCD00000475
InChI Key UUNGXYVLXYGZKH-UHFFFAOYSA-M
Synonym butylmagnesium chloride, magnesium, butylchloro, nbumgcl, bumgcl, butylmagnesiumchloride, n-butylmagnesium chloride, n-butyl-magnesium chloride, butylmagnesium chloride solution, butylmagnesium chloride solution, 2.0 m in diethyl ether, unii-4j83fgw852
PubChem CID 101837
SMILES CCCC[Mg]Cl

Specifications

Color Brown to Brown-Green
CAS Min % 19.0, 78.0, 78.0
CAS Max % 22.0, 81.0, 81.0
Physical Form Solution
Quantity 800 mL
Linear Formula CH3(CH2)3MgCl
Flash Point −21°C
IUPAC Name butyl(chloro)magnesium
Formula Weight 116.87
Percent Purity 19 to 22% (as BUMC)
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1
Density 1.0000g/mL
Chemical Name or Material n-Butylmagnesium chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.