missing translation for 'onlineSavingsMsg'
Learn More

n-Butyltin trichloride, 96%

Product Code. 11304819
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11304819 100 g 100g
11314819 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11304819 Supplier Thermo Scientific Alfa Aesar Supplier No. 071125.22
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1118-46-3 | C4H9Cl3Sn | 282.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1118-46-3
Molecular Formula C4H9Cl3Sn
Molecular Weight (g/mol) 282.176
MDL Number MFCD00000515
InChI Key YMLFYGFCXGNERH-UHFFFAOYSA-K
Synonym butyltin trichloride, stannane, butyltrichloro, monobutyltin trichloride, butyltrichlorotin, trichlorobutyltin, butyl trichloro stannane, n-butyltin trichloride, trichlorobutylstannane, stannane, trichlorobutyl, monotributyltin trichloride
PubChem CID 14236
IUPAC Name butyl(trichloro)stannane
SMILES CCCC[Sn](Cl)(Cl)Cl

Specifications

Melting Point -63°C
Density 1.693 g/mL
Boiling Point 98°C (10 mmHg)
Flash Point 81°C (178°F)
Odor Acrid
Refractive Index 1.5244
Quantity 100 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 282.17
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material n-Butyltin trichloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.