missing translation for 'onlineSavingsMsg'
Learn More

n-Butylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 15495315
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495315 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15495315 Supplier Thermo Scientific Alfa Aesar Supplier No. H58121.AD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 92273-73-9 | C4H9BrZn | 202.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Reacts with water.

Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with oxidizing agents, acid chlorides. Protect from humidity and water. Do not together with acids.
TRUSTED_SUSTAINABILITY

Specifications

CAS 92273-73-9
Density 0.958
Molecular Formula C4H9BrZn
Linear Formula CH3(CH2)3ZnBr
MDL Number MFCD00671997
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym bromo butyl zinc, n-butylzinc bromide, buznbr, butyl zinc bromide, n-buznbr, n-butyl zinc bromide, butylzinc bromide solution, hmbgxqklghidmn-uhfffaoysa-m, butylzinc bromide, 0.50 m in thf, butylzinc bromide solution, 0.5 m in thf
Solubility Information Reacts with water.
InChI Key PRWJKDARXFAPOY-UHFFFAOYSA-M
SMILES CCC[CH2-].[Zn+]Br
IUPAC Name bromozinc(1+);butane
Molecular Weight (g/mol) 202.4
PubChem CID 5225082
Formula Weight 202.4
Chemical Name or Material n-Butylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Butylzinc bromide, 0.5M in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
Supplemental information
  • MIXTURE LIST-Contain: Tetrahydrofuran, n-Butylzinc bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.