missing translation for 'onlineSavingsMsg'
Learn More

n-Butyric acid, 99+%

Product Code. 11904931 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
100mL
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11904931 10 L Plastic drum 10 L
10294320 100 mL Glass bottle 100mL
10090880 1 L Glass bottle 1 L
10428060 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Sea freight only: For EU stock allow up to 15 days delivery.
Codice prodotto. 11904931 Fornitore Thermo Scientific Acros N. del fornitore 108110100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 107-92-6 | C4H8O2 | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 107-92-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002814
InChI Key FERIUCNNQQJTOY-UHFFFAOYSA-N
Synonym butyric acid, n-butyric acid, n-butanoic acid, propylformic acid, ethylacetic acid, butanic acid, 1-propanecarboxylic acid, butyrate, 1-butyric acid, buttersaeure
PubChem CID 264
ChEBI CHEBI:30772
IUPAC Name butanoic acid
SMILES CCCC(O)=O

Specifica

Melting Point -7.0°C to +5.0°C
Boiling Point 162.0°C to 165.0°C
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula CH3CH2CH2CO2H
UN Number 2820
Solubility Information Solubility in water: miscible. Other solubilities: miscible with almost all common organic solvents
Formula Weight 88.11
Percent Purity ≥99%
Density 0.9600g/mL
Quantity 10 L
Flash Point 69°C
Infrared Spectrum Authentic
Packaging Plastic drum
Physical Form Liquid
Refractive Index 1.3965 to 1.3995
Specific Gravity 0.96
Chemical Name or Material n-Butyric Acid
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.