missing translation for 'onlineSavingsMsg'
Learn More

N-Carbamoyl-DL-aspartic acid, 98%

Product Code. 11315069
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11315069 50 g 50g
11305069 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11315069 Supplier Thermo Scientific Alfa Aesar Supplier No. A17166.18
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 923-37-5 | C5H8N2O5 | 176.128 g/mol

N-Carbamoyl-DL-aspartic acid, is a useful biochemical for proteomics research. It is also used as a carbamate derivative which serves as an intermediate in pyrimidine biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Carbamoyl-DL-aspartic acid, is a useful biochemical for proteomics research. It is also used as a carbamate derivative which serves as an intermediate in pyrimidine biosynthesis.

Solubility
Soluble in hot water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 923-37-5
Molecular Formula C5H8N2O5
Molecular Weight (g/mol) 176.128
MDL Number MFCD00042822
InChI Key HLKXYZVTANABHZ-UHFFFAOYSA-N
Synonym ureidosuccinic acid, 2-ureidosuccinic acid, n-carbamoyl-dl-aspartic acid, n-aminocarbonyl-dl-aspartic acid, dl-aspartic acid, n-aminocarbonyl, 2-carbamoylamino butanedioic acid, n-carbamoylaspartic acid, carbamoyl-dl-aspartic acid, carbamylaspartic acid, 2-ureidobutanedioic acid
PubChem CID 279
ChEBI CHEBI:64850
IUPAC Name 2-(carbamoylamino)butanedioic acid
SMILES C(C(C(=O)O)NC(=O)N)C(=O)O

Specifications

Melting Point ∼175°C (decomposition)
Beilstein 1726861
Solubility Information Soluble in hot water.
Quantity 50 g
Formula Weight 176.13
Percent Purity 98%
Chemical Name or Material N-Carbamoyl-DL-aspartic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.