missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Cyclohexylaniline, 98%

Product Code. 11469583
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469583 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11469583 Supplier Alfa Aesar™ Supplier No. A12409.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
N-Cyclohexylaniline is used as reagent in palladium-catalyzed amination of aromatic bromides. It is also used to produce N-cyclohexyl-N-phenyl-urea at ambient temperature.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1821-36-9
Melting Point 14°C to 15°C
Boiling Point 279°C
Flash Point 136°C (277°F)
Molecular Formula C12H17N
MDL Number MFCD00014284
Quantity 10 g
Beilstein 2209864
Synonym n-phenylcyclohexylamine, benzenamine, n-cyclohexyl, aniline, n-cyclohexyl, cyclohexanamine, n-phenyl, diphenylamine, ar-hexahydro, cyclohexylamine, n-phenyl, cyclohexylphenylamine, n1-cyclohexylaniline, cyclohexyl-phenyl-amine, phenylcyclohexylamine
InChI Key TXTHKGMZDDTZFD-UHFFFAOYSA-N
SMILES C1CCC(CC1)NC2=CC=CC=C2
IUPAC Name N-cyclohexylaniline
Molecular Weight (g/mol) 175.275
PubChem CID 74567
Formula Weight 175.28
Percent Purity 98%
Chemical Name or Material N-Cyclohexylaniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.