missing translation for 'onlineSavingsMsg'
Learn More

N-Cyclohexylethanolamine, 97%, Thermo Scientific™

Product Code. 11446770
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446770 100 g 100g
11436770 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446770 Supplier Thermo Scientific Alfa Aesar Supplier No. L10821.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2842-38-8
Molecular Formula C8H17NO
Molecular Weight (g/mol) 143.23
MDL Number MFCD00021326
InChI Key MGUMZJAQENFQKN-UHFFFAOYSA-N
Synonym n-cyclohexylethanolamine, 2-cyclohexylamino ethanol, ethanol, 2-cyclohexylamino, abbomeen e-25 aerosol, cyclohexlaminoethanol, abromeen e-25, 2-cyclohexylamino ethan-1-ol, n-2-hydroxyethyl cyclohexylamine, cyclohexylamine, hydroxyethyl
PubChem CID 17827
IUPAC Name 2-(cyclohexylamino)ethanol
SMILES C1CCC(CC1)NCCO

Specifications

Melting Point 33°C to 38°C
Boiling Point 112°C (3 mmHg)
Quantity 100 g
Beilstein 1071413
Formula Weight 143.23
Percent Purity 97%
Chemical Name or Material N-Cyclohexylethanolamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.