missing translation for 'onlineSavingsMsg'
Learn More

N-Cyclohexylhydroxylamine hydrochloride, 98%

Product Code. 11352216
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352216 5 g 5g
11342216 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352216 Supplier Thermo Scientific Alfa Aesar Supplier No. L03569.06
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25100-12-3 | C6H14ClNO | 151.634 g/mol

N-Cyclohexylhydroxylamine hydrochloride is used in the preparation of 4-carbethoxy-2-cyclohexyl-5-(2H)-isoxazolone and hexahydroisoxazolequinoline analogs. It is also used in the preparation of N-cyclohexyl nitrone by reaction with aldehyde. Further, it acts as a hydroxylamination reagent for allyl esters. In addition to this, it serves as a reagent in the conversion of 1,2-dicarboxylic acids to alfa-beta-unsaturated carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Cyclohexylhydroxylamine hydrochloride is used in the preparation of 4-carbethoxy-2-cyclohexyl-5-(2H)-isoxazolone and hexahydroisoxazolequinoline analogs. It is also used in the preparation of N-cyclohexyl nitrone by reaction with aldehyde. Further, it acts as a hydroxylamination reagent for allyl esters. In addition to this, it serves as a reagent in the conversion of 1,2-dicarboxylic acids to alfa-beta-unsaturated carboxylic acids.

Solubility
Soluble in water.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25100-12-3
Molecular Formula C6H14ClNO
Molecular Weight (g/mol) 151.634
MDL Number MFCD00012565
InChI Key SSVAHXZUFFSFER-UHFFFAOYSA-N
Synonym n-cyclohexylhydroxylamine hydrochloride, cyclohexylhydroxylamine hydrochloride, unii-r98vsp3t5l, r98vsp3t5l, cyclohexylhydroxylamine hcl, acmc-1ch2g, n-cyclohexylhydroxylarnine hydrochloride, n-cyclohexyl hydroxylamine hydrochloride, cyclohexanamine,n-hydroxy-,hydrochloride 1:1
PubChem CID 3084432
IUPAC Name N-cyclohexylhydroxylamine;hydrochloride
SMILES C1CCC(CC1)NO.Cl

Specifications

Melting Point ∽166°C (decomposition)
Quantity 5 g
Beilstein 3672798
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 151.64
Percent Purity 98%
Chemical Name or Material N-Cyclohexylhydroxylamine hydrochloride
compliance-icons
Product Identifier
  • N-Cyclohexylhydroxylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Cyclohexylhydroxylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.