missing translation for 'onlineSavingsMsg'
Learn More

N-Cyclohexylsulfamic Acid(Cyclamic Acid), TRC

Product Code. 30180053
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180053 10 g 10g
30179866 50 mg 50mg
30179830 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180053 Supplier Toronto Research Chemicals Supplier No. TRCC98772010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-88-9
Molecular Formula C6 H13 N O3 S
Molecular Weight (g/mol) 179.24
Synonym Cyclamic acid, Sulfamic acid, N-cyclohexyl-, Cyclohexanesulfamic acid (6CI,7CI,8CI), Sulfamic acid, cyclohexyl- (9CI), N-Cyclohexylsulfamic acid, Cyclamate, Cyclohexylamidosulfuric acid, Cyclohexylaminesulfonic acid, Cyclohexylsulfamic acid, Hexamic acid, NSC 220327, Polycat 200
IUPAC Name cyclohexylsulfamic acid
SMILES OS(=O)(=O)NC1CCCCC1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H13NO3S/c8-11(9,10)7-6-4-2-1-3-5-6/h6-7H,1-5H2,(H,8,9,10)
Formula Weight 179.0616
Chemical Name or Material Cyclamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.