missing translation for 'onlineSavingsMsg'
Learn More

N-De[2-(methylsulfonyl)ethyl] Lapatinib, TRC

Product Code. 30364700
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364700 5 mg 5mg
30363805 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30364700 Supplier Toronto Research Chemicals Supplier No. TRCD2312705MG
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 697299-82-4
Molecular Formula C26H20ClFN4O2
Molecular Weight (g/mol) 474.91
Synonym 6-[5-(Aminomethyl)-2-furanyl]-N-[3-chloro-4-[(3-fluorophenyl)methoxy]phenyl]-4-quinazolinamine, [6-(5-Aminomethylfuran-2-yl)quinazolin-4-yl][3-chloro- 4-(3-fluorobenzyloxy)phenyl]amine
SMILES Cc1nc2c3c(ccc2nc1)n(C)c(N)n3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C26H20ClFN4O2/c27-22-12-19(5-8-25(22)33-14-16-2-1-3-18(28)10-16)32-26-21-11-17(4-7-23(21)30-15-31-26)24-9-6-20(13-29)34-24/h1-12,15H,13-14,29H2,(H,30,31,32)
Chemical Name or Material N-De[2-(methylsulfonyl)ethyl] Lapatinib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.